Learn More
Strem, An Ascensus Company CAS# 7440-05-3. 1g. Palladium black (99.9%). MFCD00011167
SDP
Catalog No. 5090112389
Encompass
CAS# 7440-05-3. 1g. Palladium black (99.9%). MFCD00011167. Molecular Weight: 106.40. Molecular Formula: Pd. Color/form: pwdr. Strem# 46-1830. http://www.strem.com/catalog/v/46-1830/
Catalog No. 50-901-12389 Supplier Strem, An Ascensus Company Supplier No. 4618301G
CAS# 7440-05-3. 1g. Palladium black (99.9%). MFCD00011167. Molecular Weight: 106.40. Molecular Formula: Pd. Color/form: pwdr. Strem# 46-1830. http://www.strem.com/catalog/v/46-1830/