Learn More
Strem, An Ascensus Company CAS# 614753-51-4. 250MG. Chloro(di-2-norbornylphosphino)(2-dimethylaminomethylferrocen-1-yl)palladium(II), min. 97%. MFCD05861622
SDP
Supplier:  Strem, An Ascensus Company 460272250MG
Encompass
CAS# 614753-51-4. 250MG. Chloro(di-2-norbornylphosphino)(2-dimethylaminomethylferrocen-1-yl)palladium(II), min. 97%. MFCD05861622. Molecular Formula: C27H39ClFeNPPd. Molecular Weight: 606.31. Color/Form: beige pwdr. Strem# 46-0272. www.strem.com/catalog/v3/46-0272/
Catalog No. 50-196-3595