Learn More
Arbor Assays Palladium PdX API Fluorescent Kit (1 Plate)
SDP
Catalog No. 502278389
Encompass_Preferred
The DetectX Palladium API Screening Fluorescent Detection Kit is designed to allow rapid screening of stepwise API methods for Pd removal in a variety of solvents and includes a reference standard and proprietary PdXtm Palladium Detection Reagent. 
This item is not returnable. View return policy
Catalog No. 50-227-8389 Supplier Arbor Assays Supplier No. K007F1
The DetectX Palladium API Screening Fluorescent Detection Kit is designed to allow rapid screening of stepwise API methods for Pd removal in a variety of solvents and includes a reference standard and proprietary PdXtm Palladium Detection Reagent. 