Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palladium hydroxide | 12135-22-7 | MFCD00064599 | >97.0% | 140.43 g/mol | Pd(OH)2 | 5 G
SDP

Catalog No. 5000209878
Encompass_Preferred

Palladium hydroxide on activated carbon (20% loading, wetted with approximately 55% water) is a supported palladium catalyst supplied as a wet solid for laboratory hydrogenation and reduction reactions. It is typically supplied in small gram quantities and is intended for research use.

  • 20% palladium hydroxide on activated carbon, support wetted with approximately 55% water.
  • Used as a heterogeneous hydrogenation and reduction catalyst.
  • Form: wet solid, light brown to black, suitable for small-scale laboratory use.
  • Purity: ≥97.0%.
  • CAS number: 12135-22-7; chemical formula: Pd(OH)2; molecular weight: 140.43 g/mol.
  • Storage: store at room temperature; in solvent: -80°C for 2 years, -20°C for 1 year.

Catalog No. 50-002-09878 Supplier Medchemexpress LLC Supplier No. HYW0484575G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palladium hydroxide on activated carbon (20% loading, wetted with approximately 55% water) is a supported palladium catalyst supplied as a wet solid for laboratory hydrogenation and reduction reactions. It is typically supplied in small gram quantities and is intended for research use.

  • 20% palladium hydroxide on activated carbon, support wetted with approximately 55% water.
  • Used as a heterogeneous hydrogenation and reduction catalyst.
  • Form: wet solid, light brown to black, suitable for small-scale laboratory use.
  • Purity: ≥97.0%.
  • CAS number: 12135-22-7; chemical formula: Pd(OH)2; molecular weight: 140.43 g/mol.
  • Storage: store at room temperature; in solvent: -80°C for 2 years, -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.