Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palladium hydroxide on activated carbon | 12135-22-7 | ≥97.0% | Pd(OH)2 (supported on activated carbon) | 10 G
SDP

Catalog No. 5000210346
Encompass_Preferred

Palladium hydroxide on activated carbon is a supported palladium catalyst supplied as a wet solid (approximately 55% water). It contains 20% palladium hydroxide dispersed on activated carbon and is intended for laboratory research use such as hydrogenation and deprotection reactions.

  • 20% palladium hydroxide on activated carbon
  • Wetted with ca. 55% water
  • Purity ≥97.0%
  • Appearance light brown to black solid
  • Used as a hydrogenation and deprotection catalyst in laboratory research

Catalog No. 50-002-10346 Supplier Medchemexpress LLC Supplier No. HYW04845710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palladium hydroxide on activated carbon is a supported palladium catalyst supplied as a wet solid (approximately 55% water). It contains 20% palladium hydroxide dispersed on activated carbon and is intended for laboratory research use such as hydrogenation and deprotection reactions.

  • 20% palladium hydroxide on activated carbon
  • Wetted with ca. 55% water
  • Purity ≥97.0%
  • Appearance light brown to black solid
  • Used as a hydrogenation and deprotection catalyst in laboratory research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.