Learn More
Medchemexpress LLC Palladium hydroxide on activated carbon | 12135-22-7 | MFCD00064599 | >=97.0% | 140.43 | Pd(OH)2 | 25 G
SDP
Catalog No. 5000209148
Encompass_Preferred
Palladium hydroxide on activated carbon (20%) wetted with about 55% water is a heterogeneous palladium catalyst used for laboratory hydrogenation and hydrogenolysis. The material consists of palladium hydroxide supported on activated carbon with ~20% palladium loading by weight and is supplied as a wet solid for ease of handling.
  • 20% palladium loading on activated carbon.
  • Wetted form contains approximately 55% water by weight.
  • Solid appearance: light brown to black.
  • High assay purity (≥97.0%).
  • Supplied in small pack sizes suitable for laboratory use (e.g., 25 G).
  • Store at room temperature; typical shelf life listed as 3 years.
Catalog No. 50-002-09148 Supplier Medchemexpress LLC Supplier No. 25GHYW048457
Palladium hydroxide on activated carbon (20%) wetted with about 55% water is a heterogeneous palladium catalyst used for laboratory hydrogenation and hydrogenolysis. The material consists of palladium hydroxide supported on activated carbon with ~20% palladium loading by weight and is supplied as a wet solid for ease of handling.
  • 20% palladium loading on activated carbon.
  • Wetted form contains approximately 55% water by weight.
  • Solid appearance: light brown to black.
  • High assay purity (≥97.0%).
  • Supplied in small pack sizes suitable for laboratory use (e.g., 25 G).
  • Store at room temperature; typical shelf life listed as 3 years.