Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palladium (II) acetate | 3375-31-3 | 224.51 | 10 G
SDP

Catalog No. 5000447523
Encompass_Preferred

Palladium (II) acetate is a catalyst that catalyzes aromatic substitution reactions. It is a yellow to brown solid, intended for research use only.

  • Catalyst for aromatic substitution reactions
  • For research use only
  • Shipped at room temperature in the continental US
  • Store at 4°C, protected from light, and under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protected from light, stored under nitrogen)
  • Solubility of 100 mg/mL in DMSO (445.41 mM)
  • Recommended use of freshly opened DMSO due to hygroscopic properties
  • Appearance: Yellow to brown solid

Catalog No. 50-004-47523 Supplier Medchemexpress LLC Supplier No. HYW00207210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palladium (II) acetate is a catalyst that catalyzes aromatic substitution reactions. It is a yellow to brown solid, intended for research use only.

  • Catalyst for aromatic substitution reactions
  • For research use only
  • Shipped at room temperature in the continental US
  • Store at 4°C, protected from light, and under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (protected from light, stored under nitrogen)
  • Solubility of 100 mg/mL in DMSO (445.41 mM)
  • Recommended use of freshly opened DMSO due to hygroscopic properties
  • Appearance: Yellow to brown solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.