Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palladium (II) acetate | 3375-31-3 | 224.51 | 5 G
SDP

Catalog No. 5000447692
Encompass_Preferred

Palladium (II) acetate is a catalyst that catalyzes aromatic substitution reactions.

  • Appearance: Solid
  • Color: Yellow to brown
  • Molecular weight: 224.51
  • Purity: ≥99.0%
  • Storage: 4°C, protect from light, stored under nitrogen
  • In solvent storage: -80°C, 6 months; -20°C, 1 month (protect from light, stored under nitrogen)
  • Solubility in DMSO: 100 mg/mL (445.41 mM)
  • Solubility in Vivo Protocol 1: ≥ 2.5 mg/mL (11.14 mM)

Catalog No. 50-004-47692 Supplier Medchemexpress LLC Supplier No. HYW0020725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palladium (II) acetate is a catalyst that catalyzes aromatic substitution reactions.

  • Appearance: Solid
  • Color: Yellow to brown
  • Molecular weight: 224.51
  • Purity: ≥99.0%
  • Storage: 4°C, protect from light, stored under nitrogen
  • In solvent storage: -80°C, 6 months; -20°C, 1 month (protect from light, stored under nitrogen)
  • Solubility in DMSO: 100 mg/mL (445.41 mM)
  • Solubility in Vivo Protocol 1: ≥ 2.5 mg/mL (11.14 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.