Learn More
Strem, An Ascensus Company CAS# 3375-31-3.Palladium(II) acetate, 99+% (99.95+%-Pd). MFCD00012453. 1G.
SDP
Catalog No. 5090112384
Encompass
CAS# 3375-31-3. 1G. Palladium(II) acetate, 99+% (99.95+%-Pd). MFCD00012453. Molecular Formula: C4H6O4Pd. Molecular Weight: 224.5. Color/Form: golden-brown xtl. Strem#: 46-1781 www.strem.com/catalog/v3/46-1781/
Catalog No. 50-901-12384 Supplier Strem, An Ascensus Company Supplier No. 4617811G
CAS# 3375-31-3. 1G. Palladium(II) acetate, 99+% (99.95+%-Pd). MFCD00012453. Molecular Formula: C4H6O4Pd. Molecular Weight: 224.5. Color/Form: golden-brown xtl. Strem#: 46-1781 www.strem.com/catalog/v3/46-1781/