Learn More
Strem, An Ascensus Company CAS# 3375-31-3. 1g. Palladium(II) acetate, min. 98% (99.9+%-Pd). MFCD00012453
SDP
Catalog No. 5090112381
Encompass
CAS# 3375-31-3. 1g. Palladium(II) acetate, min. 98% (99.9+%-Pd). MFCD00012453. Molecular Weight: 224.5. Molecular Formula: Pd(OOCCH3)2. Color/form: golden-brown xtl. Strem# 46-1780. http://www.strem.com/catalog/v/46-1780/
Catalog No. 50-901-12381 Supplier Strem, An Ascensus Company Supplier No. 4617801G
CAS# 3375-31-3. 1g. Palladium(II) acetate, min. 98% (99.9+%-Pd). MFCD00012453. Molecular Weight: 224.5. Molecular Formula: Pd(OOCCH3)2. Color/form: golden-brown xtl. Strem# 46-1780. http://www.strem.com/catalog/v/46-1780/