Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) acetate, Pd 45.9-48.4%

Catalog No. AA1051606
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1051606 5 g
AA1051603 1 g
AA1051614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1051606 Supplier Thermo Scientific Chemicals Supplier No. 01051606
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3375-31-3 | C4H6O4Pd | 224.51 g/mol

Palladium(II)acetate is a catalyst of choice for a wide variety of reactions such as vinylation, Wacker process, Buchwald-Hartwig amination, carbonylation, oxidation, rearrangement of dienes (e.g., Cope rearrangement), C-C bond formation, reductive amination, etc. Precursor to Pd(0), other Pd(II) compounds of catalytic significance, and Pd nanowires.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3375-31-3
Molecular Formula C4H6O4Pd
Molecular Weight (g/mol) 224.51
MDL Number MFCD00012453
InChI Key YJVFFLUZDVXJQI-UHFFFAOYSA-L
Synonym palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
PubChem CID 167845
IUPAC Name palladium(2+);diacetate
SMILES [Pd++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point ∽205°C (decomposition)
Color Red to Brown
Physical Form Needles
Quantity 5 g
Assay Percent Range 45.9 to 48.4%
Solubility Information Soluble as monomer in glacial acetic acid or as trimer in benzene.
Formula Weight 673.46
Appearance Various forms in red-brown color (powder/flake/crystalline/beads)
Chemical Name or Material Palladium(II) acetate
Hazard Category H290-H318
Hazard Statement Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P234-P280-P305+P351+P338-P310-P390
EINECSNumber 222-164-4
RTECSNumber AJ9000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.