Learn More
Palladium(II) acetate, Pd 45.9-48.4%
Description
Palladium(II)acetate is a catalyst of choice for a wide variety of reactions such as vinylation, Wacker process, Buchwald-Hartwig amination, carbonylation, oxidation, rearrangement of dienes (e.g., Cope rearrangement), C-C bond formation, reductive amination, etc. Precursor to Pd(0), other Pd(II) compounds of catalytic significance, and Pd nanowires.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∽205°C (decomposition) |
| Color | Red to Brown |
| Physical Form | Needles |
| Quantity | 25 g |
| Appearance | Various forms in red-brown color (powder/flake/crystalline/beads) |
| Assay Percent Range | 45.9 to 48.4% |
| Solubility Information | Soluble as monomer in glacial acetic acid or as trimer in benzene. |
| Formula Weight | 673.46 |
| Chemical Name or Material | Palladium(II) acetate |
Safety and Handling
| Hazard Category | H290-H318 |
| Hazard Statement | Avoid contact with skin. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P234-P280-P305+P351+P338-P310-P390 |
| EINECSNumber | 222-164-4 |
| RTECSNumber | AJ9000000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.