Learn More
Medchemexpress LLC Palladium(II) acetylacetonate | 14024-61-4 | MFCD00000025 | 98.0% | 304.62 g·mol-1 | C10H14O4Pd | 250 MG
SDP
Catalog No. 5000302821
Encompass_Preferred
Palladium(II) acetylacetonate is a palladium coordination compound commonly used as a catalyst precursor in organic synthesis and materials chemistry. It is used in homogeneous catalysis and C-C coupling reactions and is supplied as a high-purity reagent for research applications.
  • Used as a catalyst precursor in organic synthesis.
  • Suitable for C-C coupling and other palladium-catalyzed transformations.
  • High chemical purity for research applications.
  • Solid coordination complex, convenient to handle and store.
Catalog No. 50-003-02821 Supplier Medchemexpress LLC Supplier No. 250MGHYW020952
Palladium(II) acetylacetonate is a palladium coordination compound commonly used as a catalyst precursor in organic synthesis and materials chemistry. It is used in homogeneous catalysis and C-C coupling reactions and is supplied as a high-purity reagent for research applications.
  • Used as a catalyst precursor in organic synthesis.
  • Suitable for C-C coupling and other palladium-catalyzed transformations.
  • High chemical purity for research applications.
  • Solid coordination complex, convenient to handle and store.