Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium(II) bromide

Catalog No. 2058772G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-587-72G 2 g
20-587-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-587-72G Supplier Sigma Aldrich Supplier No. 2058772G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium(II) bromide (PdBr2) is widely used as Pd(II) catalyst. PdBr2 can be prepared by reacting palladium metal with HNO3 and HBr. Electrosynthesis of PdBr2 from palladium metal and HBr has been reported.Palladium(II) bromide (PdBr2) has been used as a source of Pd(0) precatalyst in various coupling reactions. It was also employed as catalyst for the carbonylative Heck coupling reaction of aryl bromides and vinyl ethers.

Specifications

CAS 13444-94-5
Quantity 2 g
Molecular Formula Br2Pd
Linear Formula Br2Pd
MDL Number MFCD00011170
Synonym Palladium dibromide; Palladous bromide
Molecular Weight (g/mol) 266.23
Percent Purity 99%
Density 5.173 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.