Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) bromide, Premion™, 99.99% (metals basis), Pd 39.5% min

Catalog No. AA1187809
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1187809 10 g
AA1187804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1187809 Supplier Thermo Scientific Chemicals Supplier No. 01187809
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13444-94-5 | H2O2Pd | 140.43 g/mol

Palladium(II) bromide is used as an oxidizing agent, as Pd(II) catalyst and Pd(0) precatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-94-5
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
IUPAC Name palladium(2+) dihydroxide
SMILES [OH-].[OH-].[Pd++]

Specifications

Melting Point (decomposition)
Color Black
Physical Form Powder
Quantity 10 g
Odor Odorless
Sensitivity Hygroscopic
Appearance Black moist powder
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 99.99% (metal basis)
Solubility Information soluble in hydrogen bromide. Insoluble in water.
Formula Weight 266.22
Chemical Name or Material Palladium hydroxide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-588-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.