Learn More
Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min
Description
Palladium(II) bromide is used as a source of the palladium(0) precatalyst in various coupling reactions. It is used as a catalyst for the carbonylative Heck coupling reaction. It is also used as an oxidizing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | Black |
Physical Form | Powder |
Quantity | 1 g |
Solubility Information | Insoluble in water. Soluble in hydrogen bromide. |
Formula Weight | 266.22 |
Percent Purity | 99.998% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Pd 39.5% min |
Assay | (metals basis) |
Show More |
Safety and Handling
missing translation for 'healthHazard1' | H315-H319-H335 |
missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 236-588-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.