Learn More
Strem, An Ascensus Company CAS# 7647-10-1. 5g. Palladium(II) chloride (99.9%-Pd). MFCD00003558
SDP
Catalog No. 5090112394
Encompass
CAS# 7647-10-1. 5g. Palladium(II) chloride (99.9%-Pd). MFCD00003558. Molecular Weight: 177.31. Molecular Formula: PdCl2. Color/form: rust colored xtl. Strem# 46-1850. http://www.strem.com/catalog/v/46-1850/
Catalog No. 50-901-12394 Supplier Strem, An Ascensus Company Supplier No. 4618505G
CAS# 7647-10-1. 5g. Palladium(II) chloride (99.9%-Pd). MFCD00003558. Molecular Weight: 177.31. Molecular Formula: PdCl2. Color/form: rust colored xtl. Strem# 46-1850. http://www.strem.com/catalog/v/46-1850/