Learn More
Strem, An Ascensus Company CAS# 64916-48-9. 5g. Palladium(II) hexafluoroacetylacetonate, min. 95%. MFCD00015709
SDP
Catalog No. 5081513
Encompass
CAS# 64916-48-9. 5g. Palladium(II) hexafluoroacetylacetonate, min. 95%. MFCD00015709. Molecular Weight: 520.52. Molecular Formula: Pd(CF3COCHCOCF3)2. Color/form: yellow pwdr. Strem# 46-1870. http://www.strem.com/catalog/v/46-1870/
Catalog No. 50-815-13 Supplier Strem, An Ascensus Company Supplier No. 4618705G
CAS# 64916-48-9. 5g. Palladium(II) hexafluoroacetylacetonate, min. 95%. MFCD00015709. Molecular Weight: 520.52. Molecular Formula: Pd(CF3COCHCOCF3)2. Color/form: yellow pwdr. Strem# 46-1870. http://www.strem.com/catalog/v/46-1870/