Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium(II) hexafluoroacetylacetonate

Catalog No. 4014711G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-147-11G 1 g
40-147-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-147-11G Supplier Sigma Aldrich Supplier No. 4014711G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in a redox transmetalation reaction to enhance CVD alloying technology for copper. Also used in the simultaneous etching of copper and deposition of palladium for applications in ultra large-scale integrated devices.

Specifications

CAS 64916-48-9
Molecular Formula C10H2F12O4Pd
Linear Formula Pd(C5HF6O2)2
Quantity 1 g
MDL Number MFCD00015709
Synonym Pd(II) hexafluoroacetylacetonate
Molecular Weight (g/mol) 520.52
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.