Learn More
Strem, An Ascensus Company CAS# 7790-38-7. 5g. Palladium(II) iodide, 99%. MFCD00011171
SDP
Catalog No. 5090115367
Encompass
CAS# 7790-38-7. 5g. Palladium(II) iodide, 99%. MFCD00011171. Molecular Weight: 360.23. Molecular Formula: PdI2. Color/form: black xtl. Strem# 93-4607. http://www.strem.com/catalog/v/93-4607/
Catalog No. 50-901-15367 Supplier Strem, An Ascensus Company Supplier No. 9346075G
CAS# 7790-38-7. 5g. Palladium(II) iodide, 99%. MFCD00011171. Molecular Weight: 360.23. Molecular Formula: PdI2. Color/form: black xtl. Strem# 93-4607. http://www.strem.com/catalog/v/93-4607/