Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) iodide, 99.9% (metals basis), Pd 28% min

Catalog No. AA1188104
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1188104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1188104 Supplier Thermo Scientific Chemicals Supplier No. 01188104
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7790-38-7 | I2Pd | 360.23 g/mol

Palladium(II) iodide is used in gravimetric analysis. It is involved in the synthesis of symmetrical and unsymmetrical ureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-38-7
Molecular Formula I2Pd
Molecular Weight (g/mol) 360.23
MDL Number MFCD00011171
InChI Key HNNUTDROYPGBMR-UHFFFAOYSA-L
IUPAC Name palladium(2+) diiodide
SMILES [Pd++].[I-].[I-]

Specifications

Melting Point 350°C (decomposition)
Color Black
Physical Form Powder
Quantity 2 g
Density 6.003 g/mL
Solubility Information Soluble in ammonia,methyl acetate and potassium iodide. Slightly soluble in hydriodic acid and concentrated nitric acid. Insoluble in water,ethanol,diluted nitric acid,hydrochloric acid and diethyl ether.
Formula Weight 360.23
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 28% min.
Assay (metals basis)
Chemical Name or Material Palladium(II) iodide
Show More Show Less
Hazard Category H315-H319-H334
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P280-P285-P302+P352-P304+P341-P305+P351+P338-P332+P313-P342+P311-P362-P501c
EINECSNumber 232-203-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.