Learn More
Strem, An Ascensus Company CAS# 10102-05-3. 5g. Palladium(II) nitrate hydrate (Pd ~40%) (99.9%-Pd). MFCD00149818
SDP
Catalog No. 5090115369
Encompass
CAS# 10102-05-3. 5g. Palladium(II) nitrate hydrate (Pd ~40%) (99.9%-Pd). MFCD00149818. Molecular Weight: 230.72. Molecular Formula: Pd(NO3)2∙XH2O. Color/form: brown xtl. Strem# 93-4608. http://www.strem.com/catalog/v/93-4608/
Catalog No. 50-901-15369 Supplier Strem, An Ascensus Company Supplier No. 9346085G
CAS# 10102-05-3. 5g. Palladium(II) nitrate hydrate (Pd ~40%) (99.9%-Pd). MFCD00149818. Molecular Weight: 230.72. Molecular Formula: Pd(NO3)2∙XH2O. Color/form: brown xtl. Strem# 93-4608. http://www.strem.com/catalog/v/93-4608/