Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium(II) Nitrate Hydrate

Catalog No. 20576110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-576-110G 10 g
20-576-12G 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 20-576-110G Supplier Sigma Aldrich Supplier No. 20576110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Preparation of platinum-palladium/carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cellsSynthesis of Cu-Pd alloy thin films on Ti substrates by co-electrodeposition of Pd and Cu from nitrate-base electrolytic bathsPreparation of palladium catalyst supported on vertically aligned multi-walled carbon nanotubes for microwave-assisted Heck reactions of p-iodonitrobenzene with styrene and Et acrylatePreparation of a sinter-resistant Pd/SiO2 nanocatalystPreparation of di-phenyl sulfide-modified Pd/TiO2 catalysts for acetylene hydrogenationSynthesis of alumina-supported palladium catalysts active for the gas phase hydrogenation of crotonaldehyde

Specifications

CAS 207596-32-5
Quantity 10 g
Molecular Formula N2O6Pd -+ xH2O
Linear Formula Pd(NO3)2-+xH2O
MDL Number MFCD00149818
Synonym Palladium dinitrate hydrate; Palladium nitrate hydrate; Palladous nitrate hydrate
Molecular Weight (g/mol) 230.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.