Learn More
Strem, An Ascensus Company CAS# 42196-31-6. 5g. Palladium(II) trifluoroacetate, min. 97%. MFCD00013204
SDP
Catalog No. 5090112321
Encompass
CAS# 42196-31-6. 5g. Palladium(II) trifluoroacetate, min. 97%. MFCD00013204. Molecular Weight: 332.44. Molecular Formula: Pd(OOCCF3)2. Color/form: brown pwdr. Strem# 46-0280. http://www.strem.com/catalog/v/46-0280/
Catalog No. 50-901-12321 Supplier Strem, An Ascensus Company Supplier No. 4602805G
CAS# 42196-31-6. 5g. Palladium(II) trifluoroacetate, min. 97%. MFCD00013204. Molecular Weight: 332.44. Molecular Formula: Pd(OOCCF3)2. Color/form: brown pwdr. Strem# 46-0280. http://www.strem.com/catalog/v/46-0280/