Learn More
Palladium Nickel gauze, 50 mesh woven from 0.1578mm (0.0062in) dia wire
Description
Palladium Nickel gauze is used in filtrations. It is also used as a capture gauze for platinum recovery in ammonia oxidation apparatus with a pressure of 4 atm (405 kPa) and a temperature of 8500C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Palladium Nickel gauze |
| Name Note | 50 mesh woven from 0.1578mm (0.0062 in.) dia. wire |
| Avg. Mol. Wt. or Mol. Wt. Range | Pd:Ni; 95:5 wt% |
| Quantity | 150 x 150 mm |
| Mesh Size | 50 Mesh |
| Form | Wire Cloth |
| MDL Number | MFCD05863544 |
| Solubility Information | Insoluble in water. |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.