Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution, Specpure™

Catalog No. AA45291AK
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA45291AK 250 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45291AK Supplier Thermo Scientific Chemicals Supplier No. 045291AK
Add to Cart

missing translation for 'validMainframeMsg'

0.1% Pd and 0.06% Mg(NO3)2 in 2% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 0.1% Pd and 0.06% Mg(NO3)2 in 2% HNO3
MDL Number MFCD09910543

Specifications

Packaging Plastic bottle
Quantity 250 mL
UN Number UN3264
Physical Form Liquid
Chemical Name or Material Palladium nitrate & Magnesium nitrate
Hazard Category H290-H315-H319
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.