Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium On Activated Charcoal

Catalog No. 7599050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-990-50G 50 g
75-990-10G 10 g
75-990-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 75-990-50G Supplier Sigma Aldrich Supplier No. 7599050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.Palladium on activated charcoal has been used for the reduction of trimesic acid to form a mixture of cis,cis and cis,trans isomers of 1,3,5-cyclohexanetricarboxylic acid. It can also be used for the conversion of benzylic and allylic alcohols into the corresponding carbonyl compounds in the presence of olefins as hydrogen acceptors.Catalyst for dehydrogenations

Specifications

Linear Formula Pd
MDL Number MFCD03457879
Quantity 50 g
Synonym Pd/C; Pd/C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.