Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium On Barium Sulfate

Catalog No. 20572950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-572-950G 50 g
20-572-910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 20-572-950G Supplier Sigma Aldrich Supplier No. 20572950G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium on barium sulfate (Pd/BaSO4), also known as Rosenmund catalyst, is used for the decarboxylation of aliphatic esters for further production of biodiesel. It can also be used in hydrogenation reaction. Pd/BaSO4 may be used as a catalyst reservoir for the Stille cross-coupling reaction between iodobenzene and tributylphenyltin.We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Specifications

Linear Formula Pd
MDL Number MFCD03613605
Quantity 50 g
Synonym Pd/BaSO4; Rosenmund catalyst
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.