Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium On Carbon

Catalog No. 20567225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-567-225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-567-225G Supplier Sigma Aldrich Supplier No. 20567225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.Palladium on carbon (Pd/C) (1 wt. % loading, matrix activated carbon support) can be used for a variety of applications such asa catalyst for hydrodechlorination of dichlorodifluoromethane (CCl2F2), which is a known greenhouse gasa catalyst for Heck reactions of aryl halides in ionic liquidsan anode catalyst for the fabrication of direct formic acid fuel cells (DFAFC)a cost-effective electrocatalyst for the oxidation of methanolin the catalytic hydrogenation of chemical hydrides for the production of hydrogena catalyst in Suzuki-Mayaura reaction

Specifications

CAS 13139-12-3
Molecular Formula C8H17NO3
Linear Formula C8H17NO3
MDL Number MFCD00037766
Quantity 5 g
Molecular Weight (g/mol) 175.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.