Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium Silver foil, 0.025mm (0.001in) thick, 99.9% (metals basis excluding Pt)

Catalog No. AA42187GH
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42187GH 100 x 100 mm
AA42187FF 25 x 25 mm
AA42187FI 50 x 50 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42187GH Supplier Thermo Scientific Chemicals Supplier No. 042187GH
Add to Cart

missing translation for 'validMainframeMsg'

Pd:Ag; 75:25 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Pd:Ag; 75:25 wt%
MDL Number MFCD02091738

Specifications

Packaging Tube
Quantity 100 x 100 mm
Percent Purity 99.9%
Chemical Name or Material Palladium Silver foil
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.