Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palladium trifluoroacetate | 42196-31-6 | MFCD00013204 | 332.45 | 250 MG
SDP

Catalog No. 5000450038
Encompass_Preferred

Palladium trifluoroacetate is a biochemical reagent used in life science research. It appears as a light brown to gray solid and is soluble in DMSO. This product requires sealed storage away from moisture, with specific conditions for storage in solvent.

  • Biochemical reagent
  • Suitable for life science research
  • Light brown to gray solid
  • Soluble in DMSO (50 mg/mL)
  • Store at 4°C, sealed, away from moisture
  • In solvent, stable for 6 months at -80°C
  • In solvent, stable for 1 month at -20°C

Catalog No. 50-004-50038 Supplier Medchemexpress LLC Supplier No. 250MGHYW007333
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palladium trifluoroacetate is a biochemical reagent used in life science research. It appears as a light brown to gray solid and is soluble in DMSO. This product requires sealed storage away from moisture, with specific conditions for storage in solvent.

  • Biochemical reagent
  • Suitable for life science research
  • Light brown to gray solid
  • Soluble in DMSO (50 mg/mL)
  • Store at 4°C, sealed, away from moisture
  • In solvent, stable for 6 months at -80°C
  • In solvent, stable for 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.