Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advanced Instruments Palladox™ Disposable Sachet
SDP

Catalog No. 22047608 Shop All Advanced Instruments Products
Change view
Click to view available options
Quantity:
40 Sachets/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-047-608 40 Sachets/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-047-608 Supplier Advanced Instruments Supplier No. AN3146
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Advanced Instruments Palladox™ Disposable Sachet is compatible with all anaerobic jar sizes

  • The Palladox™ Disposable Sachet is designed specifically to work with the Anoxomat™ Anaerobic Jar System
  • Simplifies anaerobic sample testing by minimizing catalyst handling time
  • Reduces the oxygen in the jar within minutes
  • Maintains an anaerobic environment for the duration of incubation
  • No oven required for regeneration
  • Compatible with all jar sizes
  • One year shelf life

Specifications

Type Single Use Catalyst
Content And Storage 36°F to 72°F, 0 to 50% RH
Dimensions 5 x 3.75 x 5.75 in.
Disposable Single-use
For Use With (Application) Oxygen Reduction
For Use With (Equipment) Anoxomat™ Anaerobic Jar System
Includes 40 Sachets/ Pack
Quantity 40 Sachets/Pk.
Weight (English) 0.625 lb.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.