Learn More
Saf T Pak Inc Imperium Plus Half Pallet Insulated Shipping Solution
SDP
Catalog No. 501904586 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
Inmark's Imperium family of patent-pending pallet shippers provides the performance and flexibility to solve your bulk distribution challenges.Effectively manage and mitigate risk by tuning to your organization's specific risk profile(s), cost targets and business objectives.FEATURES: Performs to ISTA 7D Standard: Rigorously designed and developed to ISTA 7D Standard, the Imperium family of pallet shippers ensure performance in the chamber is only exceeded by performance in the field.One Shared Platform - 18 Configurations: The Imperium family incorporates 3 sizes with 18 potential configurations all built on the same platform with shared common parts allowing for built-to-fit payload configurations.Variable Temperature System: Imperium shipping systems are capable of meeting numerous temperature and duration requirements by a simple configuration change.
Catalog No. 50-190-4586 Supplier Saf T Pak Inc Supplier No. 37664
Inmark's Imperium family of patent-pending pallet shippers provides the performance and flexibility to solve your bulk distribution challenges.Effectively manage and mitigate risk by tuning to your organization's specific risk profile(s), cost targets and business objectives.FEATURES: Performs to ISTA 7D Standard: Rigorously designed and developed to ISTA 7D Standard, the Imperium family of pallet shippers ensure performance in the chamber is only exceeded by performance in the field.One Shared Platform - 18 Configurations: The Imperium family incorporates 3 sizes with 18 potential configurations all built on the same platform with shared common parts allowing for built-to-fit payload configurations.Variable Temperature System: Imperium shipping systems are capable of meeting numerous temperature and duration requirements by a simple configuration change.