Learn More
Grainger Pinnacle Floor Scale, Carbon Steel, 5,000 lb Capacity, 1 ea
Catalog No. 502779419
Encompass_Preferred
This digital platform scale is a heavy-duty floor scale designed for warehouse and industrial weighing applications. It features a 5,000 lb (2,268 kg) capacity with a 1 lb (0.5 kg) readability. Constructed with durable steel and a two-part epoxy paint finish, the system includes a 48 in. x 48 in. diamond tread platform and a remote digital indicator with a 1.2 in. LED display.
  • 5,000 lb (2,268 kg) capacity with 1 lb (0.5 kg) readability
  • 48 in. x 48 in. heavy-duty steel platform with diamond tread
  • NTEP Class III rated for legal-for-trade applications
  • Equipped with four alloy steel shear beam load cells
  • RS-232 communications port for optional printer connectivity
  • Top access junction box and adjustable feet for simplified maintenance
  • Includes 20 ft. armored interface cable with quick connect
  • Designed for 120 VAC power operation
Catalog No. 50-277-9419 Supplier Grainger Supplier No. 19ND56
This digital platform scale is a heavy-duty floor scale designed for warehouse and industrial weighing applications. It features a 5,000 lb (2,268 kg) capacity with a 1 lb (0.5 kg) readability. Constructed with durable steel and a two-part epoxy paint finish, the system includes a 48 in. x 48 in. diamond tread platform and a remote digital indicator with a 1.2 in. LED display.
  • 5,000 lb (2,268 kg) capacity with 1 lb (0.5 kg) readability
  • 48 in. x 48 in. heavy-duty steel platform with diamond tread
  • NTEP Class III rated for legal-for-trade applications
  • Equipped with four alloy steel shear beam load cells
  • RS-232 communications port for optional printer connectivity
  • Top access junction box and adjustable feet for simplified maintenance
  • Includes 20 ft. armored interface cable with quick connect
  • Designed for 120 VAC power operation