Learn More
Uline Pallet Rack Wire Decking 46x48""""
Catalog No. NC1920900
Encompass
Welded steel wire decking for pallet racks, 46 x 48"""", 2,500 pound capacity, waterfall edge, 3 support channels, galvanized 2.5 x 4"""" mesh, 6 gauge wire
Catalog No. NC1920900 Supplier Uline Supplier No. H5725
Welded steel wire decking for pallet racks, 46 x 48"""", 2,500 pound capacity, waterfall edge, 3 support channels, galvanized 2.5 x 4"""" mesh, 6 gauge wire