Learn More
American Radiolabeled Chemicals Inc PALM AC 16-3H 1LC
SDP
Catalog No. 50352972
Encompass_Preferred
Palmitic acid-16-3H 1mCi in ethanol, 1 mCi/ml, 20-30 Ci/mmol, M.W. 256.4, Exclusive
Catalog No. 50-352-972 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 19861MC
Palmitic acid-16-3H 1mCi in ethanol, 1 mCi/ml, 20-30 Ci/mmol, M.W. 256.4, Exclusive