Learn More
Aviva Systems Biology Corp PALM Antibody - N-terminal region (ARP56270_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56270_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PALM. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PALM. Alternate names: KIAA0270ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-134