Learn More
Cole-Parmer Extech PH220-C Palm pH Meter with Cabled Electrode
Catalog No. 50992409 Shop All Cole-Parmer Products
Encompass
Big Digit LCD display in rugged housing with choice of stick or cabled pH ElectrodeLarge LCD displays pH or mV and temperature simultaneouslyMicroprocessor provides for automatic buffer recognitionMemory stores 15 readingsAutomatic temperature compensation via Pt-100 sensor built into electrodeUnit is completely waterproof when electrode is
Catalog No. 50-992-409 Supplier Cole-Parmer Supplier No. 5302643
Big Digit LCD display in rugged housing with choice of stick or cabled pH ElectrodeLarge LCD displays pH or mV and temperature simultaneouslyMicroprocessor provides for automatic buffer recognitionMemory stores 15 readingsAutomatic temperature compensation via Pt-100 sensor built into electrodeUnit is completely waterproof when electrode is