Learn More
Description
- Commonly used in law enforcement and forensic applications to support identification alongside fingerprints.
- Made from durable 110 lb. cardstock, they work with standard fingerprint ink.
- Compatible with any type of fingerprinting ink
- Printed on both front and back
Specifications
Specifications
| Dimensions (L x W) | 8 x 8 in. |
| Product Type | Palm Print Record Card |
| For Use With (Application) | Palm Printing |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
