Learn More
Cayman Chemical Palmarumycin C3, Purity: ≥95%, CAS: 159934-11-9, 2.5 mg
SDP
Catalog No. 502991057
Encompass_Preferred
A fungal metabolite; active against A. tumefaciens, B. subtilis, P. lachrymans, R. solanacearum, S. haemolyticus, and X. vesicatoria (MICs = 6.25, 6.25, 12.5, 12.5, 6.25, and 6.25 µg/ml, respectively); active against M. microspora, E. repens, F. oxysporum, and U. violacea in agar diffusion assays at 0.5 mg/disc; scavenges DPPH radicals in cell-free assays and has antioxidant activity in a β-carotene-linoleic acid bleaching assay (IC50s = 37.57 and 7.41 µg/ml, respectively)
Catalog No. 50-299-1057 Supplier Cayman Chemical Supplier No. 314472.5 MG
A fungal metabolite; active against A. tumefaciens, B. subtilis, P. lachrymans, R. solanacearum, S. haemolyticus, and X. vesicatoria (MICs = 6.25, 6.25, 12.5, 12.5, 6.25, and 6.25 µg/ml, respectively); active against M. microspora, E. repens, F. oxysporum, and U. violacea in agar diffusion assays at 0.5 mg/disc; scavenges DPPH radicals in cell-free assays and has antioxidant activity in a β-carotene-linoleic acid bleaching assay (IC50s = 37.57 and 7.41 µg/ml, respectively)