Learn More
Selleck Chemical LLC Palmatine 25mg 3486-67-7 Berbericinine, Burasaine
SDP
Supplier:  Selleck Chemical LLC S376925MG
Encompass_Preferred
Palmatine (Berbericinine, Burasaine), an isoquinoline alkaloid, has sedative, antidepressant, antioxidative, anti-ulcerative, antacid, anticancer, and anti-metastatic activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8409