Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmatine hydroxide | 131-04-4 | 99.6% | 369.41 | 1 G
SDP

Catalog No. 5000428560
Encompass_Preferred

Palmatine hydroxide | 131-04-4 | 99.6% | 369.41 | 1 G

Catalog No. 50-004-28560 Supplier Medchemexpress LLC Supplier No. HYN0110B1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmatine hydroxide | 131-04-4 | 99.6% | 369.41 | 1 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.