Learn More
Medchemexpress LLC Palmatine hydroxide | 131-04-4 | 99.6% | 369.41 | 250 MG
SDP
Catalog No. 5000430472
Encompass_Preferred
Palmatine hydroxide is an orally active, irreversible indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor. It also inhibits West Nile virus (WNV) NS2B-NS3 protease in an uncompetitive manner.
  • Orally active, irreversible IDO-1 inhibitor
  • Inhibits WNV NS2B-NS3 protease
  • Shows anti-cancer activities
  • Possesses anti-oxidation properties
  • Exhibits anti-inflammatory effects
  • Provides neuroprotection
  • Has antibacterial activities
  • Demonstrates anti-viral activities
Catalog No. 50-004-30472 Supplier Medchemexpress LLC Supplier No. 250MGHYN0110B
Palmatine hydroxide is an orally active, irreversible indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor. It also inhibits West Nile virus (WNV) NS2B-NS3 protease in an uncompetitive manner.
  • Orally active, irreversible IDO-1 inhibitor
  • Inhibits WNV NS2B-NS3 protease
  • Shows anti-cancer activities
  • Possesses anti-oxidation properties
  • Exhibits anti-inflammatory effects
  • Provides neuroprotection
  • Has antibacterial activities
  • Demonstrates anti-viral activities