Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmaturbine hydroxide | 5 MG
SDP

Catalog No. 5000421076
Encompass_Preferred

Palmaturbine hydroxide is a natural product isolated from *Tinospora sinensis*. It is classified as an alkaloid, specifically an isoquinoline alkaloid, and also as a phenol and monophenol. This compound is provided as a solid with a purity of ≥99.0% and has documented solubility in DMSO.

  • Isolated from *Tinospora sinensis*
  • Purity ≥99.0%
  • Soluble in DMSO (100 mg/mL)
  • Recommended storage at 4°C, protected from light

Catalog No. 50-004-21076 Supplier Medchemexpress LLC Supplier No. HYN2373A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmaturbine hydroxide is a natural product isolated from *Tinospora sinensis*. It is classified as an alkaloid, specifically an isoquinoline alkaloid, and also as a phenol and monophenol. This compound is provided as a solid with a purity of ≥99.0% and has documented solubility in DMSO.

  • Isolated from *Tinospora sinensis*
  • Purity ≥99.0%
  • Soluble in DMSO (100 mg/mL)
  • Recommended storage at 4°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.