Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmaturbine hydroxide | 98.3% | 355.38 g/mol | C20H21NO5 | 10 MG
SDP

Catalog No. 5000288931
Encompass_Preferred

Palmaturbine hydroxide is a naturally derived alkaloid supplied for laboratory research use. Provided as the hydroxide salt in milligram-scale vials, it is accompanied by manufacturer documentation (data sheet, certificate of analysis, and safety data sheet). This material is for research use only and not for human or clinical applications.

  • Natural alkaloid hydroxide for research use
  • Purity 98.32%
  • Chemical formula C20H21NO5; molecular weight 355.38 g/mol
  • Available in small milligram pack sizes (5 mg, 10 mg, 50 mg, 100 mg)
  • Store at 4°C and protect from light; in solution: -80°C (up to 6 months) or -20°C (up to 1 month)
  • Supplied with data sheet, COA, and SDS

Catalog No. 50-002-88931 Supplier Medchemexpress LLC Supplier No. HYN2373A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmaturbine hydroxide is a naturally derived alkaloid supplied for laboratory research use. Provided as the hydroxide salt in milligram-scale vials, it is accompanied by manufacturer documentation (data sheet, certificate of analysis, and safety data sheet). This material is for research use only and not for human or clinical applications.

  • Natural alkaloid hydroxide for research use
  • Purity 98.32%
  • Chemical formula C20H21NO5; molecular weight 355.38 g/mol
  • Available in small milligram pack sizes (5 mg, 10 mg, 50 mg, 100 mg)
  • Store at 4°C and protect from light; in solution: -80°C (up to 6 months) or -20°C (up to 1 month)
  • Supplied with data sheet, COA, and SDS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.