Learn More
Cayman Chemical Palmitate (calcium salt), Purity: >98%, CAS: 542-42-7, 100 mg
SDP
Catalog No. 502887566
Encompass_Preferred
A neutral, lipid soluble form of the free acid; increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol
Catalog No. 50-288-7566 Supplier Cayman Chemical Supplier No. 10010279100 MG
A neutral, lipid soluble form of the free acid; increases cytosolic Ca2+ concentration leading to pancreatic acinar cell injury due to excessive consumption of ethanol