Learn More
Medchemexpress LLC Palmitelaidic Acid | 10030-73-6 | 25 MG
SDP
Catalog No. 5000420662
Encompass_Preferred
Palmitelaidic acid (9-trans-Hexadecenoic acid) is the trans isomer of palmitoleic acid, a monounsaturated fatty acid abundant in serum and tissue. It is correlated with cardiometabolic risk factors and promotes glucose uptake while improving insulin sensitivity. This substance down-regulates proinflammatory adipocytokine genes and lipogenic genes in the liver.
  • Trans isomer of palmitoleic acid
  • Promotes faster glucose uptake
  • Increases phosphorylation of AMPK
  • Up-regulates glucokinase
  • Down-regulates SREBP-1
  • Increases production of FGF-21
  • Stimulates expression of PPARα
  • Reduces body weight increase
  • Ameliorates development of hyperglycemia and hypertriglyceridemia
  • Improves insulin sensitivity
  • Down-regulates proinflammatory adipocytokine genes (TNFα and resistin)
  • Down-regulates lipogenic genes (SREBP-1, FAS, and SCD-1) in the liver
Catalog No. 50-004-20662 Supplier Medchemexpress LLC Supplier No. 25MGHYN2341
Palmitelaidic acid (9-trans-Hexadecenoic acid) is the trans isomer of palmitoleic acid, a monounsaturated fatty acid abundant in serum and tissue. It is correlated with cardiometabolic risk factors and promotes glucose uptake while improving insulin sensitivity. This substance down-regulates proinflammatory adipocytokine genes and lipogenic genes in the liver.
  • Trans isomer of palmitoleic acid
  • Promotes faster glucose uptake
  • Increases phosphorylation of AMPK
  • Up-regulates glucokinase
  • Down-regulates SREBP-1
  • Increases production of FGF-21
  • Stimulates expression of PPARα
  • Reduces body weight increase
  • Ameliorates development of hyperglycemia and hypertriglyceridemia
  • Improves insulin sensitivity
  • Down-regulates proinflammatory adipocytokine genes (TNFα and resistin)
  • Down-regulates lipogenic genes (SREBP-1, FAS, and SCD-1) in the liver