Learn More
Cayman Chemical Palmitelaidic Acid, Purity: ≥98%, CAS: 10030-73-6, 500 mg
SDP
Catalog No. 502892957
Encompass_Preferred
A dietary fatty acid; decreases NO production and levels of soluble E-selectin in isolated HAECs at 20 µM; inhibits Sendai virus-induced hemolysis of isolated human erythrocytes at 49 µM; increased serum levels are associated with higher levels of LDL, but lower levels of triglycerides, fasting insulin, blood pressure, and incidence of diabetes
Catalog No. 50-289-2957 Supplier Cayman Chemical Supplier No. 9001798500 MG
A dietary fatty acid; decreases NO production and levels of soluble E-selectin in isolated HAECs at 20 µM; inhibits Sendai virus-induced hemolysis of isolated human erythrocytes at 49 µM; increased serum levels are associated with higher levels of LDL, but lower levels of triglycerides, fasting insulin, blood pressure, and incidence of diabetes