Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Palmitelaidic acid (9-trans-hexadecenoic acid) | 10030-73-6 | MFCD00067230 | 99.4% | 254.41 g/mol | C16H30O2 | 10 MG
SDP

Catalog No. 5000289154
Encompass_Preferred

Palmitelaidic acid (9-trans-hexadecenoic acid) is the trans isomer of palmitoleic acid, supplied as a high-purity research reagent for biochemical and lipid studies.

  • High purity: 99.4%.
  • Chemical identifier: CAS 10030-73-6.
  • Chemical formula: C16H30O2; molecular weight: 254.41 g/mol.
  • Available sizes: 10 MG, 25 MG, 50 MG.
  • Typical formulation for 10 MG: 393 mM in 100 μL ethanol.
  • Application: used in fatty acid metabolism studies and biochemical assays.

Catalog No. 50-002-89154 Supplier Medchemexpress LLC Supplier No. HYN234110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Palmitelaidic acid (9-trans-hexadecenoic acid) is the trans isomer of palmitoleic acid, supplied as a high-purity research reagent for biochemical and lipid studies.

  • High purity: 99.4%.
  • Chemical identifier: CAS 10030-73-6.
  • Chemical formula: C16H30O2; molecular weight: 254.41 g/mol.
  • Available sizes: 10 MG, 25 MG, 50 MG.
  • Typical formulation for 10 MG: 393 mM in 100 μL ethanol.
  • Application: used in fatty acid metabolism studies and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.