Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palmitoleic acid, >99%, MP Biomedicals™

Catalog No. ICN15604480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
0.5 g
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15604480 100 mg
ICN15604490 0.5 g
ICN15604491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15604480 Supplier MP Biomedicals, Inc Supplier No. 0215604480
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61-25-6
Molecular Formula C20H22ClNO4
Molecular Weight (g/mol) 375.849
InChI Key UOTMYNBWXDUBNX-UHFFFAOYSA-N
Synonym papaverine hydrochloride, cardiospan, cardoverina, dispamil, drapavel, forpavin, papalease, papaversan, pavatest, paverolan
PubChem CID 6084
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxyisoquinoline;hydrochloride
SMILES COC1=C(C=C(C=C1)CC2=NC=CC3=CC(=C(C=C32)OC)OC)OC.Cl

Specifications

Color Colorless
Density 0.895g/mL at 20°C
Assay Percent Range ≥99%
Quantity 100 mg
Formula Weight 254.4
Percent Purity >99%
Physical Form Frozen Liquid
Chemical Name or Material Palmitoleic Acid
EINECSNumber 206-765-9
Recommended Storage Store at -0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.